missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YSI™ Pro Series DO Cap Membrane Kit
Specifications
Specifications
| Color | Yellow |
| Type | Membrane Kit |
| For Use With (Application) | For pro series galvanic sensors |
| Includes | Bottle of NaCl electrolyte, 1 sanding disc, six screw-on cap 1.25mL Polyethylene make DO membranes, yellow color |
| Material | Polyethylene Membrane |