Learn More
YSI™ MultiLab Line Benchtop IDS Conductivity Sensors
Sensors maintain their calibration data and prevent recalibration
Supplier:  YSI™ 301711Y
 View more versions of this product
Catalog No. 09324556
Only null left
Description
Description
  • Intelligent digital sensor for measuring conductivity
  • Intelligent digital sensor store their own unique ID with serial number and calibration data
  • Plug and play connectivity with all MultiLab instruments
  • Automatic sensor recognition with digital signal processing and data transfer
Warranty
Two years
Specifications
Chemical Identifiers
0.2°C
120 mm
0°C to 100°C
4.7 in.
32°F to 212°F
Specifications
0.2°C
15.3 mm
120 mm
3 m
0 to 14
0 to 1999g/L
0°C to 100°C
0.6 in.
4.7 in.
118.1 in.
Epoxy
0 to 70ppt
32°F to 212°F
Product Suggestions
Product Suggestions