Learn More
YSI™ Cap Membrane Kits
Description
Includes six cap membranes, a 1 oz. (30mL) bottle of KCl solution and one sanding disk. For use with 13-398-87, -88, and -89 (Model 5239 Field Oxygen/Temperature Probes); 13-299-76 (Model 5905 BOD Probe); and 13-298-24 through-27 (Model 85 DO/Conductivity Meters).
Specifications
Specifications
| For Use With (Equipment) | Field Oxygen/Temperature Probe, BOD Probe, DO/Conductivity Meter |
| Type | Cap Memebrane Kit |
| Includes | Six cap membranes, a 1 oz. (30 mL) bottle of KCl solution and one sanding disk |
| Quantity | 1 Kit |