Learn More
YSI™ 3220 Temperature Probe for 3200 Conductivity Instruments
Catalog No. 0932442
Change view
Click to view available options
Probe Type:
Temperature
Catalog No. Probe Type
0932442 Temperature
1 options
Catalog No. 0932442 Supplier YSI™ Supplier No. 3220
Use with conductivity cells that lack a built-in temperature sensor on YSI 3200 Conductivity instruments
  • For automatic temperature compensation
Specifications
For Use With (Application) For automatic temperature compensation
Probe Type Temperature
For Use With (Equipment) Conductance meter