missing translation for 'onlineSavingsMsg'
Learn More

Youngstown Barrel & Drum Tight Head Steel Drum

Catalog No. 18930A
Change view
Click to view available options
Capacity (Metric):
18.92 L
114 L
208.19 L
Certifications/Compliance:
UN 1A1/X1.2/300-DOT/UN
UN 1A1/Y1.8/100-DOT/UN
UN 1A1/Y1.8/250-DOT/UN
Product Type:
Drum
Lined Drum
Pail
Unlined Drum
Material:
Carbon Steel
Steel
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Certifications/Compliance Product Type Material
18930A 114 L UN 1A1/Y1.8/250-DOT/UN Unlined Drum Carbon Steel
18929C 208.19 L UN 1A1/X1.2/300-DOT/UN Unlined Drum Carbon Steel
18931C 18.92 L UN 1A1/Y1.8/100-DOT/UN Pail Steel
191304687 114 L UN 1A1/Y1.8/250-DOT/UN Lined Drum Carbon Steel
191304848 208.19 L UN 1A1/X1.2/300-DOT/UN Lined Drum Carbon Steel
19171252 208.19 L UN 1A1/X1.2/300-DOT/UN Drum Carbon Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18930A Supplier Youngstown Barrel & Drum Supplier No. 457

Youngstown Tight Head Steel Pail is perfect for the transporting of hazardous solids or liquids.

With cold-rolled carbon-steel construction, steel drums are qualified for safe transport of noncorrosive hazardous liquids or solids.

  • Salvage drum designed to overpack and ship damaged and/or leaking drums
  • Also qualify for direct containment of hazardous solids

Specifications

Certifications/Compliance UN 1A1/Y1.8/250-DOT/UN
Product Type Unlined Drum
Closure Type Bung
Description 20 Gauge
Capacity (Metric) 114 L
Color Black
For Use With (Application) For transport of hazardous solids or liquids, Designed to overpack and ship damaged and/or leaking drums
Material Carbon Steel
Freight Class 50
Head Style Tight
Quantity Ea.
Height (English) 29 in.
Height (Metric) 73.66 cm
Diameter (English) Outer 18.25 in.
Diameter (Metric) Outer 46.35 mm
Weight (English) 24 lb.
Weight (Metric) 10.88 Kg
Show More Show Less