missing translation for 'onlineSavingsMsg'
Learn More

Youngstown Barrel & Drum Plastic Open Head Pails with Screw Lids

Catalog No. 18120240
Change view
Click to view available options
Capacity (English):
1.25 gal.
3.5 gal.
5 gal.
6.5 gal.
Includes:
Screw Lid w/ Neoprene Gasket
Screw Lid w/ Neoprene Gasket, BIOHAZARD Label
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Includes
18120240 3.5 gal. Screw Lid w/ Neoprene Gasket, BIOHAZARD Label
18120241 6.5 gal. Screw Lid w/ Neoprene Gasket, BIOHAZARD Label
18120242 5 gal. Screw Lid w/ Neoprene Gasket, BIOHAZARD Label
19171855 6.5 gal. Screw Lid w/ Neoprene Gasket
19171856 5 gal. Screw Lid w/ Neoprene Gasket
19171857 1.25 gal. Screw Lid w/ Neoprene Gasket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18120240 Supplier Youngstown Barrel & Drum Supplier No. 2379BH
Only null left

Seamless construction prevents leaks

  • Blow-molded, high-density plastic
  • Drop handle is comfortable for carrying loaded pail
  • Handle folds down for storage or stacking
  • White

Specifications

Certifications/Compliance UN 1H2/Y19/S
Product Type Open Head Pail (Screw Lid)
Closure Type Screw Lid with Neoprene Gasket
Capacity (English) 3.5 gal.
Capacity (Metric) 13.25 L
Color White
For Use With (Application) For Packaging Small Amounts of Liquid Products
Material Plastic
Freight Class 50
Head Style Open
Includes Screw Lid w/ Neoprene Gasket, BIOHAZARD Label
Height (English) 12.19 in.
Height (Metric) 30.96 cm
Diameter (English) Inner 11.25 in.
Diameter (English) Outer 12.62 in.
Diameter (Metric) Inner 28.57 cm
Diameter (Metric) Outer 32.05 mm
Weight (English) 6.63 lb.
Weight (Metric) 3.01 Kg
Show More Show Less