missing translation for 'onlineSavingsMsg'
Learn More

Youngstown Barrel & Drum Plastic Open Head Pails with Long Skirt Lids Red

Store materials in color-coded pails

Supplier:  Youngstown Barrel & Drum YBD2400

 View more versions of this product

Catalog No. 18941


Only null left

Description

Description

Plastic Open Head Pails with short Skirt Lids. This pail is dual rated, UN 1H2/Y1.5/30 & Y30/S.

Specifications

Chemical Identifiers

Open Head Pail
18.92 L
Plastic
Open
5 gal.
For Moving and Color-coded Storage
92.5 for qty. 168

Specifications

UN 1H2/Y30/S and 1H2/Y1.5/30
Short Skirt Lid
18.92 L
For Moving and Color-coded Storage
92.5 for qty. 168
Short Skirt Lid with Gasket
30.242002 cm
36.98875 cm
11.9063 in.
30.242002 mm
1.58 Kg
Open Head Pail
5 gal.
White
Plastic
Open
30.242002 cm
14.5625 in.
11 in.
27.94 cm
3.5 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions