missing translation for 'onlineSavingsMsg'
Learn More

Yohimbine Hydrochloride 99.0+%, TCI America™

Catalog No. Y00025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Y00025G 5 g
Y000225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Y00025G Supplier TCI America Supplier No. Y00025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 65-19-0
Molecular Formula C21H27ClN2O3
Molecular Weight (g/mol) 390.91
MDL Number MFCD00012674
InChI Key PIPZGJSEDRMUAW-VJDCAHTMSA-N
Synonym yohimbine hydrochloride, antagonil, yohimbe, yohimbine hcl, aphrodine hydrochloride, yohimbine monohydrochloride, unii-nb2e1yp49f, yohimbin hydrochloride usp, nb2e1yp49f
PubChem CID 6169
IUPAC Name hydrogen methyl (1S,15R,18S,19R,20S)-18-hydroxy-3,13-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8-tetraene-19-carboxylate chloride
SMILES [H+].[Cl-].COC(=O)[C@H]1[C@@H](O)CC[C@H]2CN3CCC4=C(NC5=CC=CC=C45)[C@@H]3C[C@H]12

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1544
Formula Weight 390.91
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Yohimbine Hydrochloride
RTECSNumber ZG1015000
TSCA Yes