missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Yohimbine hydrochloride, 98%

Catalog No. p-3758990
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC141030050 5 g Glass bottle
AC141030250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC141030050 Supplier Thermo Scientific Chemicals Supplier No. 141030050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Yohimbine hydrochloride, CAS # 65-19-0, is an α2-adrenergic receptor antagonist, which blocks the central α2-adrenegic receptors in the brain. | CAS: 65-19-0 | C21H27ClN2O3 | 390.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Yohimbine hydrochloride is an indole alkaloid derived from the tree bark
  • It is an antagonist of α2-adrenergic receptors in the brain, reducing the effects of xylazine, which is an α2-adrenergic agonist

Applications

  • Yohimbine hydrochloride blocks the central α2-adrenegic receptors in the brain
  • It can reduce or eradicate effects of xylazine, which is an α2-adrenergic agonist
  • It can have an antidiuretic effect

Chemical Identifiers

CAS 65-19-0
Molecular Formula C21H27ClN2O3
Molecular Weight (g/mol) 390.91
MDL Number MFCD00012674
InChI Key PIPZGJSEDRMUAW-VJDCAHTMSA-N
Synonym yohimbine hydrochloride, antagonil, yohimbe, yohimbine hcl, aphrodine hydrochloride, yohimbine monohydrochloride, unii-nb2e1yp49f, yohimbin hydrochloride usp, nb2e1yp49f
PubChem CID 6169
IUPAC Name methyl (1S,15R,18S,19R,20S)-18-hydroxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate;hydrochloride
SMILES [H+].[Cl-].COC(=O)[C@H]1[C@@H](O)CC[C@H]2CN3CCC4=C(NC5=CC=CC=C45)[C@@H]3C[C@H]12

Specifications

Melting Point 285°C to 288°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 10300
Specific Rotation Condition +99° (20°C c=1%,H2O,dry substance)
Specific Rotation + 99
Formula Weight 390.9
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Yohimbine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-600-4

RUO – Research Use Only