missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Yohimbine hydrochloride, 98+%

CAS: 65-19-0 | C21H27ClN2O3, C21H26N2O3·HCl | 390.91 g/mol

Supplier:  Thermo Scientific Chemicals J6018503

Catalog No. AAJ6018503


Only null left

Description

Description

Yohimbine hydrochloride is an α2-adrenergic receptor antagonist. It blocks the central α2-adrenegic receptors in the brain, thus preventing and reducing the effects of xylazine, an α2-adrenergic agonist. The sedative effects and respiratory depression that accompany xylazine administration are both antagonized and reversed by yohimbine. Yohimbine hydrochloride also produces an antidiuretic effect, and increases heart rate and blood pressure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1 g
Standard alpha-2-adrenergic antagonist
65-19-0
390.91g/mol
Light Sensitive
Signal Transduction Reagent
PIPZGJSEDRMUAW-VJDCAHTMSA-N
hydrogen methyl (1S,15R,18S,19R,20S)-18-hydroxy-3,13-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8-tetraene-19-carboxylate chloride
White to Off-white
≥98%
C21H27ClN2O3
17-Hydroxyyohimban-16-carboxylic acid methyl ester hydrochloride
288°C to 290°C
Powder
[H+].[Cl-].COC(=O)[C@H]1[C@@H](O)CC[C@H]2CN3CCC4=C(NC5=CC=CC=C45)[C@@H]3C[C@H]12
390.91

Chemical Identifiers

65-19-0
390.91
17-Hydroxyyohimban-16-carboxylic acid methyl ester hydrochloride
[H+].[Cl-].COC(=O)[C@H]1[C@@H](O)CC[C@H]2CN3CCC4=C(NC5=CC=CC=C45)[C@@H]3C[C@H]12
C21H27ClN2O3
PIPZGJSEDRMUAW-VJDCAHTMSA-N
hydrogen methyl (1S,15R,18S,19R,20S)-18-hydroxy-3,13-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8-tetraene-19-carboxylate chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-600-4

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only