missing translation for 'onlineSavingsMsg'
Learn More

Yeast Extract, Powder, Spectrum™ Chemical

$207.70 - $20534.10

Specifications

Form Powder
Grade Reagent
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Type Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Type Price Quantity  
18614704
View Documents
Spectrum Chemical Mfg Cor
YE105100GM
100 g Amber Glass Bottle
Each for $207.70
Only null left
 
18614705
View Documents
Spectrum Chemical Mfg Cor
YE10512KG
12 kg Poly Pail
Each for $8,963.07
Only null left
 
18614706
View Documents
Spectrum Chemical Mfg Cor
YE10545KGBL
45 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18614707
View Documents
Spectrum Chemical Mfg Cor
YE105500GM
500 g Amber Glass Bottle
Each for $442.87
Only null left
 
Description

Description

Specifications

Specifications

Powder
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.