Learn More
MilliporeSigma™ Supelco™ Y GlasSeal™ Connector, Fused Silica
Quickly connect three pieces of fused silica tubing of the same or different diameters without the need for tools.
$725.81
Specifications
For Use With (Application) Gas Chromatography (GC)
View More Specs
Products 1
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
111008993
View Documents Promotion Details
MilliporeSigma™ Supelco™
23632
3/Pk.
Each for $725.81
Only null left
Description
Description
The leak-free connection is useful to split a sample to two columns, or to split a column effluent to two detectors. Silanized for an inert inside surface. For use with 0.10 - 0.53 mm I.D. fused silica tubing.
Specifications
Specifications
Gas Chromatography (GC)
Videos
SDS
Documents
Documents
GlasSeal is a trademark of Sigma-Aldrich Co. LLC