Learn More
Selleck Chemical Llc Y-27632 2HCl 10mg 129830-38-2 DFS Item
Supplier:  Selleck Chemicals Co Ltd S104910MG
Y-27632 2HCl is a selective ROCK1 (p160ROCK) inhibitor with Ki of 140 nM in a cell-free assay, exhibits >200-fold selectivity over other kinases, including PKC, cAMP-dependent protein kinase, MLCK and PAK. *For Research & Development use only. Product is not intended for drug, household, or human consumption.
Catalog No. 508636
Only null left
Explore available promotions