missing translation for 'onlineSavingsMsg'
Learn More

Xylylazo Violet I [Spectrophotometric reagent for Mg], TCI America™

Catalog No. A50071G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A50071G Supplier TCI America Supplier No. A50071G
Only null left

Chemical Identifiers

CAS 14936-97-1
Molecular Formula C25H20N3NaO6S
Molecular Weight (g/mol) 513.50
MDL Number MFCD00003890
InChI Key LPXSXFXFNSMAOI-MCOPDZHSSA-M
Synonym 1-Azo-2-hydroxy-3-(2,4-dimethylcarboxyanilido)naphthalene-1′C-(2-hydroxybenzene-5-sulfonic Acid) Sodium Salt, 2-[2-Hydroxy-3-(2,4-xylylcarbamoyl)-1-naphthylazo]-1-phenol-4-sulfonic Acid Sodium Salt, Xylidyl Blue I
PubChem CID 131864351
IUPAC Name sodium 3-{2-[(1Z)-3-[(2,4-dimethylphenyl)carbamoyl]-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-4-hydroxybenzene-1-sulfonate
SMILES [Na+].CC1=CC=C(NC(=O)C2=CC3=CC=CC=C3\C(=N\NC3=CC(=CC=C3O)S([O-])(=O)=O)C2=O)C(C)=C1

Specifications

Quantity 1 g
Formula Weight 513.50
Physical Form Crystalline Powder
Chemical Name or Material Xylylazo Violet I [Spectrophotometric reagent for Mg]