missing translation for 'onlineSavingsMsg'
Learn More

Xylose, USP, 98-102%, Spectrum™ Chemical

Catalog No. 18607798
Change view
Click to view available options
Quantity:
25 g
5 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607798 50 kg Fiber Drum
18607792 25 g Amber Glass Bottle
18607796 5 kg Poly Pail
18607797 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18607798 Supplier Spectrum Chemical Mfg Cor Supplier No. XY11050KGBL
Get Up to $300 in Additional Chemicals
View all promotions

missing translation for 'validMainframeMsg'

XY110, 58-86-6, C5H10O5

Spectrum™ Chemical Xylose, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 58-86-6
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key SRBFZHDQGSBBOR-UHFFFAOYNA-N
IUPAC Name oxane-2,3,4,5-tetrol
SMILES OC1COC(O)C(O)C1O

Specifications

CAS Max % 1
Ignition Residue 0.0005
Loss on Drying 0.001
Packaging Fiber Drum
Quantity 50 kg
Specific Rotation +18.2° to +19.4°
Percent Purity 98 to 102%
Grade USP