missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylobiose 98.0+%, TCI America™
$467.87 - $2311.43
Chemical Identifiers
| CAS | 6860-47-5 |
|---|---|
| Molecular Formula | C10H18O9 |
| Molecular Weight (g/mol) | 282.245 |
| MDL Number | MFCD00135984 |
| InChI Key | LGQKSQQRKHFMLI-HEMZSNRESA-N |
| Synonym | 4-O-beta-D-Xylopyranosyl-D-xylose |
| PubChem CID | 131674888 |
| IUPAC Name | (2S,3S,5R)-2-[(3R,5S)-4,5,6-trihydroxyoxan-3-yl]oxyoxane-3,4,5-triol |
| SMILES | C1C(C(C(C(O1)OC2COC(C(C2O)O)O)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
X0067100MG
|
TCI America
X0067100MG |
100 mg |
Each for $467.87
|
|
|||||
|
X00671G
|
TCI America
X00671G |
1 g |
Each for $2,311.43
|
|
|||||
Chemical Identifiers
| 6860-47-5 | |
| 282.245 | |
| LGQKSQQRKHFMLI-HEMZSNRESA-N | |
| 131674888 | |
| C1C(C(C(C(O1)OC2COC(C(C2O)O)O)O)O)O |
| C10H18O9 | |
| MFCD00135984 | |
| 4-O-beta-D-Xylopyranosyl-D-xylose | |
| (2S,3S,5R)-2-[(3R,5S)-4,5,6-trihydroxyoxan-3-yl]oxyoxane-3,4,5-triol |
Specifications
| 6860-47-5 | |
| C10H18O9 | |
| 100 mg | |
| LGQKSQQRKHFMLI-HEMZSNRESA-N | |
| (2S,3S,5R)-2-[(3R,5S)-4,5,6-trihydroxyoxan-3-yl]oxyoxane-3,4,5-triol | |
| 131674888 | |
| ≥98.0% (HPLC) | |
| Xylobiose |
| White | |
| MFCD00135984 | |
| 4-O-beta-D-Xylopyranosyl-D-xylose | |
| C1C(C(C(C(O1)OC2COC(C(C2O)O)O)O)O)O | |
| 282.245 | |
| 282.25 | |
| Crystalline Powder |
Safety and Handling
TSCA : No