missing translation for 'onlineSavingsMsg'
Learn More

Xylidine Ponceau 2R, pure

Xylidine Ponceau, CAS # 3761-53-3, is a red azo dye used in histology for staining of thin sections. | CAS: 3761-53-3 | C18H14N2Na2O7S2 | 480.417 g/mol

$149.72 - $454.73

Chemical Identifiers

CAS 3761-53-3
Molecular Formula C18H14N2Na2O7S2
Molecular Weight (g/mol) 480.417
MDL Number MFCD00003897
InChI Key YJVBLROMQZEFPA-UHFFFAOYSA-L
Synonym 4-(2,4-Dimethylphenylazo)-3-hydroxy-2,7-naphthalenedisulfonic acid, disodium salt, C.I. 16150, 4-(2, 4-Dimethylphenylazo)-3-hydroxy-2, 7-naphthalenedisulfonic acid, Ponceau G, R, 2R
PubChem CID 102092631
ChEBI CHEBI:82369
IUPAC Name disodium;5-[(2,4-dimethylphenyl)diazenyl]-6-oxido-7-sulfonaphthalene-2-sulfonate
SMILES CC1=CC(=C(C=C1)N=NC2=C3C=CC(=CC3=CC(=C2[O-])S(=O)(=O)O)S(=O)(=O)[O-])C.[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC190260250
SDS View Documents
Thermo Scientific Chemicals
190260250
25 g Glass bottle
Each for $149.72
Only null left
 
AC190261000
SDS View Documents
Thermo Scientific Chemicals
190261000
100 g Glass bottle
Each for $454.73
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Xylidine Ponceau 2R is largely confined to Masson's trichrome, where it gives a slight orange shading to the red of the cytoplasmic structures

Application

  • Xylidine Ponceau 2R is a histological dye used mainly for the staining of thin sections
  • It shows cardiotoxic and carcinogenic activities
  • It has been used to prepare Fuchsin-Ponceau solution to stain an amyotrophic lateral sclerosis tissue-engineered skin model
Specifications

Chemical Identifiers

3761-53-3
480.417
YJVBLROMQZEFPA-UHFFFAOYSA-L
102092631
disodium;5-[(2,4-dimethylphenyl)diazenyl]-6-oxido-7-sulfonaphthalene-2-sulfonate
C18H14N2Na2O7S2
MFCD00003897
4-(2,4-Dimethylphenylazo)-3-hydroxy-2,7-naphthalenedisulfonic acid, disodium salt, C.I. 16150, 4-(2, 4-Dimethylphenylazo)-3-hydroxy-2, 7-naphthalenedisulfonic acid, Ponceau G, R, 2R
CHEBI:82369
CC1=CC(=C(C=C1)N=NC2=C3C=CC(=CC3=CC(=C2[O-])S(=O)(=O)O)S(=O)(=O)[O-])C.[Na+].[Na+]

Specifications

3761-53-3
MFCD00003897
YJVBLROMQZEFPA-UHFFFAOYSA-L
CC1=CC(=C(C=C1)N=NC2=C3C=CC(=CC3=CC(=C2[O-])S(=O)(=O)O)S(=O)(=O)[O-])C.[Na+].[Na+]
480.417
CHEBI:82369
Extra Pure
Complies
25 g
Xylidine Ponceau 2R
C18H14N2Na2O7S2
4-(2,4-Dimethylphenylazo)-3-hydroxy-2,7-naphthalenedisulfonic acid, disodium salt, C.I. 16150, 4-(2, 4-Dimethylphenylazo)-3-hydroxy-2, 7-naphthalenedisulfonic acid, Ponceau G, R, 2R
Authentic
disodium;5-[(2,4-dimethylphenyl)diazenyl]-6-oxido-7-sulfonaphthalene-2-sulfonate
102092631
480.41
Glass bottle
Bordeaux to Red
Fine Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Causes serious eye irritation.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

CANCER SUSPECTED AGENT

EINECSNumber : 223-178-3

RTECSNumber : QJ6825000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only