missing translation for 'onlineSavingsMsg'
Learn More

Xylenes , Macron Fine Chemicals™

Catalog No. MK88024
Change view
Click to view available options
Quantity:
4L
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK88024 4L Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK88024 Supplier Avantor Supplier No. AVA880204

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C6H4(CH3)2

Specifications

Quantity 4L
Packaging Glass
Formula Weight 106.17
Chemical Name or Material Xylenes