Learn More
StatLab Xylene
Description
- Purified and filtered to deliver a high quality, consistent product
- High solvency factor ensures quick and complete clearing
- Free of impurities and water
- 55-Gallon Drum
Specifications
Specifications
| Product Type | Xylene |
| Quantity | 55 gal. |
| Form | Solution |