missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Xphos Pd G3
Description
Xphos Pd G3
Specifications
Specifications
| CAS | 1445085-55-1 |
| Melting Point | 146°C to 151°C |
| Molecular Formula | C46H62NO3PPdS |
| Linear Formula | C46H62NO3PPdS |
| MDL Number | MFCD22666043 |
| Quantity | 5 g |
| Synonym | (2-Dicyclohexylphosphino-2',4',6'-triisopropyl-1,1'-biphenyl)[2-(2'-amino-1,1'-biphenyl)]palladium(II) methanesulfonate; XPhos-G3-Palladacycle; XPhos-Pd-G3 |
| Molecular Weight (g/mol) | 846.45 g/mol |