missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ XLD Agar (Dehydrated)

Catalog No. OXCM0469B
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0469B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXCM0469B Supplier Thermo Scientific™ Supplier No. CM0469B
Only null left

Isolate Salmonella and Shigella from food and clinical specimens with this selective medium.

Isolate Salmonella and Shigella from clinical specimens and foods with selective Thermo Scientific™ Oxoid™ XLD Agar (Dehydrated). The medium relies on xylose fermentation, lysine decarboxylation and production of hydrogen sulphide for the primary differentiation of shigellae and salmonellae from non-pathogenic bacteria. The careful addition of sodium desoxycholate inhibits the growth of coliforms without decreasing the ability to support shigellae and salmonellae.

Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.

Use XLD Medium for the screening of samples containing mixed flora suspected of harboring enteric pathogens, e.g., medical specimens or food products.

  • Easy-to-read: easily differentiate Salmonella (red colonies with black centers) from Shigella (red colonies) and other coliforms (yellow, opaque colonies).
  • Superior sensitivity and selectivity: exceeds that of the traditional plating media e.g., Eosin Methylene Blue and Bismuth Sulphite Agars, which tend to suppress the growth of shigellae.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Description XLD Medium
Form Powder
Product Type Agar
Quantity 500 g
Yield For 9.4L medium

For Laboratory Use Only