missing translation for 'onlineSavingsMsg'
Learn More

Calcium Nitrate, Tetrahydrate, Crystal, ACS, 99-103%, Spectrum™ Chemical

Catalog No. 18609505
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609505 50 kg Poly Bottle
18609500 25 g Poly Bottle
18609501 125 g Poly Bottle
18609502 500 g Poly Bottle
18609503 2.5 kg Poly Bottle
18609504 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18609505 Supplier Spectrum Chemical Mfg Cor Supplier No. C113550KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH10N2O10
Molecular Weight (g/mol) 238.16
InChI Key DKCJOKYHYIAZQH-UHFFFAOYSA-N
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.O.O.O.[Ca++].O[N+]([O-])=O.O[N+]([O-])=O

Specifications

pH 5.0 to 7.0
Physical Form Crystalline Powder or Needles
CAS Max % 1
Quantity 50 kg
Packaging Poly Bottle
Percent Purity 99 to 103%
Grade ACS/Reagent
Chemical Name or Material Calcium Nitrate, Tetrahydrate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.