missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ XIAP Tracer mF-Smac
Description
XIAP Tracer mF-Smac is a fluorescent labeled peptide used as an acceptor dye with a terbium labeled BIR domain antibody in TR-FRET assays (Kd values are 4.7 nM at BIR3 (XIAP); 17.9 nM in XIAP polarization assay).
Specifications
Specifications
| Purity | 0.95 |
| CAS | 783340-88-5 |
| Content And Storage | Store at -20°C |
| Molecular Weight (g/mol) | 989.1 |
| Quantity | 1 mg |