missing translation for 'onlineSavingsMsg'
Learn More

Agilent Technologies™ XFp Real-time ATP Rate Assay Kit

Catalog No. 103591100
Change view
Click to view available options
Quantity:
6 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
103591100 6 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 103591100 Supplier Agilent Technologies™ Supplier No. 103591100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

XFp Real-time ATP Rate Assay Kit containing six single-use pouches, each with oligomycin and rotenone/antimycin A. For use with Seahorse XF HS Mini and XFp analyzers.

  • Simultaneously measure basal ATP production rates from mitochondrial respiration and glycolysis
  • Total ATP production rates can also be obtained
  • Assess the effect of compounds on ATP production rates
  • Generate real-time kinetic data in live cells, allowing users to assess the interplay and relationship between energy metabolism and cellular functions
  • Use with Agilent™ Seahorse XF DMEM medium, pH 7.4 or Seahorse XF RPMI medium, pH 7.4
  • Reconstitute kit components directly in the assay medium, removing the need for DMSO or other solvents
  • Eliminate the cost associated with failed experiments since each lot is rigorously tested and validated with functional assays to deliver consistent robust results
  • Upload your assay result file to Agilent™ Seahorse Analytics to calculate and report parameters of the XF Real-Time ATP Rate assay, share with collaborators, or export data

Specifications

Detection Method Fluorescence
For Use With (Equipment) Seahorse XF HS Mini Analyzer
Includes Lyophilized Reagents
Quantity 6 Assays
Type Reagent Kit