missing translation for 'onlineSavingsMsg'
Learn More

Agilent Technologies™ XF Hu T Cell Activation Assay Kit

Catalog No. 103759100
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
103759100 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 103759100 Supplier Agilent Technologies™ Supplier No. 103759100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

XF Hu T Cell Activation Assay Kit contains one vial of 2 mL ImmunoCult human CD3/CD28 T cell activator and two vials of XF 2-deoxy-D-glucose. For use with Seahorse XF HS Mini, XFp and XFe/XF96 analyzers.

  • Detects human T cell activation in minutes, providing an early window to investigate cellular and signaling requirements associated with T cell activation
  • Provides kinetic readout for continuous monitoring of human T cell activation and modulation
  • Allows dosing of cells with test compounds/modulators during assay through the built-in injection ports
  • The soluble ImmunoCult Human CD3/CD28 T Cell Activator (STEMCELL Technologies) is easy-to-use and compatible with specially-designed injection ports on Agilent™ Seahorse XF cartridges, reducing assay preparation time and improving assay reliability
  • Reagents are directly reconstituted or diluted in the assay medium, eliminating the need for DMSO or other solvents
  • Every lot is tested with functional assays to deliver consistent and robust results
  • Upload your assay result file to your Agilent™ Seahorse Analytics account to calculate and report parameters of the XF T Cell Activation assay, share with collaborators, or export data to Microsoft Excel or GraphPad Prism for other graphing or analysis needs

Specifications

Detection Method Fluorescence
For Use With (Equipment) Seahorse XF Analyzer
Quantity 1 Kit
Type Reagent Kit