Learn More
Bulbman FCR 64625 HLX Osram 54248
Catalog No. NC9141673
Encompass
Halogen Light Bulb, 12V, 100w, It is tubular with a glass diameter of 0.4 in., a GY6.35 base and an overall length of 1.7 in., It has a Kelvin temperature of 3300 and is rated at 50 life hours
Catalog No. NC9141673 Supplier Bulbman Supplier No. 104400
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Halogen Light Bulb, 12V, 100w, It is tubular with a glass diameter of 0.4 in., a GY6.35 base and an overall length of 1.7 in., It has a Kelvin temperature of 3300 and is rated at 50 life hours