Learn More
Waters Limited XBridge BEH Shield RP18 Sentry Guard Cartridge, 130, 5 m, 2.1 mm X 10 mm, 2/pk
Catalog No. 50781523
Encompass
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.
Catalog No. 50781523 Supplier Waters Limited Supplier No. 186003071
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.