Learn More
Waters Limited XBridge BEH C18 Prep Guard Cartridge, 130, 10 m, 10 mm X 10 mm, 1/pk
Catalog No. 50782048
Encompass
As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1-12). The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed.
Catalog No. 50782048 Supplier Waters Limited Supplier No. 186003889
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1-12). The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed.