missing translation for 'onlineSavingsMsg'
Learn More

Xanthydrol for the Detection of Urea, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015864
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015864 25 g
111015865 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015864 Supplier Merck Emd Millipore Supplier No. 9558025G

Chemical Identifiers

MDL Number MFCD00005057
Synonym 9-Hydroxyxanthene; 9-Xanthenol

Specifications

Melting Point 123°C to 128°C; 127°C to 128°C
CAS 90-46-0
Linear Formula C13H10O2
Quantity 25 g
Formula Weight 198.22
Percent Purity ≥99.0% (HPLC)