missing translation for 'onlineSavingsMsg'
Learn More

Xanthotoxin 98.0+%, TCI America™

Catalog No. p-7137951
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X0009100MG 100 mg
X00091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X0009100MG Supplier TCI America Supplier No. X0009100MG
Only null left

Chemical Identifiers

CAS 298-81-7
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.19
MDL Number MFCD00005009
InChI Key QXKHYNVANLEOEG-UHFFFAOYSA-N
Synonym methoxsalen, 8-methoxypsoralen, xanthotoxin, meladinine, ammoidin, oxsoralen, meloxine, oxypsoralen, xanthotoxine, meladinin
PubChem CID 4114
ChEBI CHEBI:18358
IUPAC Name 9-methoxy-7H-furo[3,2-g]chromen-7-one
SMILES COC1=C2OC=CC2=CC2=C1OC(=O)C=C2

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 100 mg
Formula Weight 216.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Xanthotoxin
EINECSNumber (9)-2281
RTECSNumber LV1400000
TSCA Yes