missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xanthosine dihydrate, 99%

Catalog No. AC207630010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC207630010 1 g
AC207630050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC207630010 Supplier Thermo Scientific Chemicals Supplier No. 207630010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5968-90-1 | C10H12N4O6·2H2O | 320.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5968-90-1
Molecular Formula C10H12N4O6·2H2O
Molecular Weight (g/mol) 320.26
InChI Key AQQAMLPKPXTPOL-GWTDSMLYSA-N
Synonym xanthosine dihydrate, unii-1u62i480sp, 9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purine-2,6-diol dihydrate, xanthosine, dihydrate, xanthosine dihydrate mi, xanthosine,dihydrate, 9-beta-d-ribofuranosyl xanthine dihydrate, 9-, a-d-ribofuranosylxanthine, xanthosine, dihydrate 8ci,9ci
PubChem CID 91886582
IUPAC Name 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purine-2,6-dione;hydrate
SMILES C1=NC2=C(N1C3C(C(C(O3)CO)O)O)NC(=O)NC2=O.O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Merck Index 14,10065
Specific Rotation Condition −53.00 (22.00°C c=8, 0.3N NaOH)
Specific Rotation -53
Formula Weight 320.26
Percent Purity 99%
Chemical Name or Material Xanthosine dihydrate

RUO – Research Use Only