missing translation for 'onlineSavingsMsg'
Learn More

Xanthopterin Hydrate 98.0+%, TCI America™

Catalog No. p-7137950
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00071G 1 g
X00075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X00071G Supplier TCI America Supplier No. X00071G

Chemical Identifiers

CAS 5979-01-1
Molecular Formula C6H7N5O3
Molecular Weight (g/mol) 197.15
MDL Number MFCD00151241
InChI Key GXYCFNCAIXIUMR-UHFFFAOYSA-N
Synonym 2-Amino-4,6-pteridinediol
PubChem CID 2723796
IUPAC Name 2-amino-1,5-dihydropteridine-4,6-dione;hydrate
SMILES C1=NC2=C(C(=O)N=C(N2)N)NC1=O.O

Specifications

Quantity 1 g
Formula Weight 179.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Xanthopterin Hydrate