missing translation for 'onlineSavingsMsg'
Learn More

Xanthone 98.0+%, TCI America™

Catalog No. X000525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X000525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X000525G Supplier TCI America Supplier No. X000525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 90-47-1
Molecular Formula C13H8O2
Molecular Weight (g/mol) 196.205
MDL Number MFCD00005060
InChI Key JNELGWHKGNBSMD-UHFFFAOYSA-N
Synonym xanthone, 9h-xanthen-9-one, 9-xanthenone, benzophenone oxide, 9-oxoxanthene, genicide, xanthenone, 9-xanthone, diphenylene ketone oxide, dibenzo-gamma-pyrone
PubChem CID 7020
ChEBI CHEBI:37647
IUPAC Name xanthen-9-one
SMILES C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3O2

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 25 g
Formula Weight 196.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Xanthone
EINECSNumber (9)-0311
RTECSNumber ZD5711000
TSCA Yes