missing translation for 'onlineSavingsMsg'
Learn More

Xanthane hydride, Thermo Scientific Chemicals

Catalog No. p-7214811
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467340050 5 g
AC467340250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467340050 Supplier Thermo Scientific Chemicals Supplier No. 467340050
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6846-35-1
Molecular Formula C2H2N2S3
Molecular Weight (g/mol) 150.23
InChI Key YWZHEXZIISFIDA-UHFFFAOYSA-N
IUPAC Name 5-amino-3H-1,2,4-dithiazole-3-thione
SMILES NC1=NC(=S)SS1

Specifications

Melting Point 145.0°C to 150.0°C
Quantity 5 g
Formula Weight 150.22
Chemical Name or Material Xanthane hydride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 229-933-3

RUO – Research Use Only