missing translation for 'onlineSavingsMsg'
Learn More

Promega φX174 DNA/HinfI Markers Conventional DNA Digest Marker

Catalog No. PRG1751
Encompass
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG1751 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG1751 Supplier Promega Supplier No. G1751

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

6X Blue/Orange Loading Dye

This conventional DNA digest marker is generated by ΦX174 DNA digested to completion with HinfI to create 20 ethanol-precipitated DNA fragments ranging in size from 24bp to 726bp.

  • Twenty DNA Fragments of 24-726bp
  • Load 1μL/lane
  • Supplied at a concentration of 1μg/μL
  • Supplied with 6X Blue/Orange Loading Dye

Specifications

Buffer 10mM Tris HCl pH 7.5, 1mM EDTA
Color Blue, Orange
Concentration 1 μg/μL
Form Liquid
Formulation 50μg X174 DNA/HinfI marker, 1mL blue/orange 6X loading dye
For Use With (Application) Separation, size determination and quantitation of DNA and RNA fragments
No. of Bands 20
Quantity 50 μg
Storage Requirements −30 to −10°C
Sufficient For 50 Lanes
Volume Per Well 1 μL
Product Type DNA Molecular Weight Marker
Show More Show Less