missing translation for 'onlineSavingsMsg'
Learn More

Corning™ X‐WASH™ System

Corning™ X‐WASH™ System is designed to wash, resuspend, and volume-reduce cell suspensions in a sterile, closed system, ensuring consistently high cell recovery and viability with short processing times.

$6,979.83 - $89,715.78

Specifications

For Use With (Equipment) X-WASH System
View More Specs

Products 3
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
10320126
View Documents Promotion Details
Corning™
6928
Control Module 1/Pk., 1/Cs. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10320127
View Documents Promotion Details
Corning™
6929
Docking Station 1/Pk., 1/Cs. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10320129
View Documents Promotion Details
Corning™
6931
Disposable Cartridges 1/Pk., 2/Cs.
Case of 20 Each for $6,979.83
Only null left
 
Description

Description

  • Process protocols can be customized to ensure consistently high cell recovery
  • Highly sensitive infrared sensors detect the interface between cells and media and direct the washed cells into a dedicated harvest chamber.
  • Harvest chamber can be pre-filled with a media of choice
    for re-suspension of harvested cells
  • Enables transfer of process data from the X-WASH Control Module to the database to support GMP processing, monitoring and reporting of information.
  • Consistent high recovery (>90%) and viability (>80%) of cells
  • Sterile, single-use, functionally closed system
  • Process volume: 40 to 240 mL
  • Average Process Time: 10 to 15 minutes/wash cycle
  • Programmable harvest volumes (3 to 40 mL)
  • Centrifugation-based cell washing system
  • Customized protocols
  • Traceability: Electronic transfer of process data
  • Bar code compatible
  • US class I medical device
Specifications

Specifications

X-WASH System
Videos
SDS
Documents

Documents