Learn More
3M™ X.Sport™ Eyewear
Ideal choice for active wear
Supplier:  3M™ 1517800000020
 View more versions of this product
Catalog No. 19-051-424
Only null left
Explore available promotions
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
Description
  • Sport-grip™ temples
  • Ratchet adjustment for three lens angles
  • 7.5 base lens is mounted in a lightweight nylon frame
  • Provides maximum coverage and excellent peripheral vision
  • Lens with DX™ anti-fog hard coat protects from fog, scratching, and most chemicals
  • 3M Catalog# 15176-00000-20 , 3M Catalog# 15182-00000-20
Compliance
Meets the requirements of ANSI™ Z87.1-2010 and CSA Z94.3-2007.
Specifications
Specifications
Meets the requirements of ANSI Z87.1-2010 and CSA Z94.3-2007.
Black
Nylon
Indoor/Outdoor Mirror
Sport Grip
Product Suggestions
Product Suggestions