Learn More
Research Products X-Ray Film, Blue Base for Autoradiography and Chemiluminescence, Non-Interleaved, 5 x 7 inch, 100 per Package
Supplier: Research Products Corporation RPI248304
RPI X-ray film has an emulsion coating on both sides of a blue tinted base. We evaluated this film in our laboratories for both autoradiography and chemiluminescence applications. Film produces high quality images with less background fog. Compatible with all intensifying screens.
Uses standard developing chemistries and can be processed manually or in an automatic processor.