Learn More
Research Products Corporation X-GAL and IPTG Ready to Use Non-Toxic Solution 10mg/ml, 100 Milligrams
Catalog No. 50488541
Encompass
Stable solution of X-GAL and IPTG. Does not contain DMSO or DMF. Solution concentration is 10 mg/ml X-Gal and 8 mg/ml IPTG.
Chromogenic substrate of β-Galactosidase used in combination with IPTG for detection of β-Galactosidase activity in bacterial colonies. X-GAL is cleaved at the B1-4 bond between galactose and the 5-Bromo-4-chloro-3-indolyl portion of X-GAL by β-Galactosidase via hydrolysis. The cleavage of X-GAL results in the production of a water insoluble blue dichloro-dibromo-indigo precipitate. In cloning strategies with vectors such as Lambda-11, M13mp18 and 19, pUC18 and 19, pUR222 the E. coli lacZ gene is transformed to lac+ cells. WARNING! This product can expose you to 1-Methyl-2-Pyrrolidon, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50488541 Supplier Research Products Corporation Supplier No. B715000.1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Stable solution of X-GAL and IPTG. Does not contain DMSO or DMF. Solution concentration is 10 mg/ml X-Gal and 8 mg/ml IPTG.
Chromogenic substrate of β-Galactosidase used in combination with IPTG for detection of β-Galactosidase activity in bacterial colonies. X-GAL is cleaved at the B1-4 bond between galactose and the 5-Bromo-4-chloro-3-indolyl portion of X-GAL by β-Galactosidase via hydrolysis. The cleavage of X-GAL results in the production of a water insoluble blue dichloro-dibromo-indigo precipitate. In cloning strategies with vectors such as Lambda-11, M13mp18 and 19, pUC18 and 19, pUR222 the E. coli lacZ gene is transformed to lac+ cells. WARNING! This product can expose you to 1-Methyl-2-Pyrrolidon, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.