Learn More
Kimberly-Clark Professional™ WypAll™ X70 Wipers
Ideal rag replacements
Supplier:  Kimberly-Clark Professional™ 06354
 View more versions of this product
Catalog No. 19137201
Only null left
Description
Description
  • Wipers combine the best qualities of disposable wipers and scrap rags
  • Super absorbent industrial cleaning wipers can be used almost anywhere to clean oil, solvents — virtually any industrial spill
  • Made from fast-absorbing Hydroknit™ material, an extra-strong nonwoven fabric blend that won't tear or fall apart even during heavy-duty cleaning
  • Textured design won't scratch delicate surfaces
Order Info
Available in quarterfold, flat, rolls and two dispensers in blue, white or red (food service).
Specifications
Chemical Identifiers
Ideal for all manufacturing and maintenance and repair
Specifications
Foodservice Towel
59.69 cm
31.75 cm
White/Red
Ideal for all manufacturing and maintenance and repair
23.5 in.
12.5 in.
Non-woven Polyproylene Cellulose
23.5 x 12.5 in.
Product Suggestions
Product Suggestions