missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark Professional™ WypAll™ X70 Wipers
Ideal rag replacements
Supplier: Kimberly-Clark Professional™ 41412
Description
- Wipers combine the best qualities of disposable wipers and scrap rags
- Super absorbent industrial cleaning wipers can be used almost anywhere to clean oil, solvents — virtually any industrial spill
- Made from fast-absorbing Hydroknit™ material, an extra-strong nonwoven fabric blend that won't tear or fall apart even during heavy-duty cleaning
- Textured design won't scratch delicate surfaces
Order Info
Available in quarterfold, flat, rolls and two dispensers in blue, white or red (food service).
Chemical Identifiers
| Ideal for all manufacturing and maintenance and repair |
Specifications
| Medium-duty Cloth | |
| 42.67 cm | |
| 23.11 cm | |
| Super | |
| Single-use | |
| Ideal for all manufacturing and maintenance and repair |
| 16.8 in. | |
| 16.8 in. | |
| Non-woven Polyproylene Cellulose | |
| Blue | |
| 16.8 x 9.1 in. |