Learn More
Bel-Art™ Write-On™ Paper Label Tapes
Use pencil, ball-point pen, or solvent ink marker to create labels that remain legible under the most demanding laboratory conditions. Paper label tapes resist moisture and most solvents and are great for use in low temperature applications.
Supplier:  Bel-Art™ F134810075
 View more versions of this product
Catalog No. 03411604
Get More Bel-Art Lab Products with Purchase
View all promotions
Only null left
Description
Description
Use pencil, ball-point pen, or solvent ink marker to create labels that remain legible under the most demanding laboratory conditions. Paper label tapes resist moisture and most solvents and are great for use in low temperature applications.
  • Adheres to any clean surface, including PTFE fluoropolymer resin coated materials
  • Peels off without leaving a sticky residue
  • Pencil, ball-point pen, or solvent ink marker remain legible under the most demanding laboratory conditions
  • Colored paper with rubber resin adhesive resists moisture and most solvents
  • Withstands a maximum temperature of 157°C (315°F) for 30 minutes or 80°C (176°F) for 24 hours; can be used down to -80°C (-112°F)
  • Withstands a steam autoclaving cycle at 121°C (250°F) and gamma irradiation between 13.0 and 45.0kGy
Specifications
Chemical Identifiers
Autoclavable
None
Roll
Ink Jet
Non-sterile
Rubber
Blank
Single-side Printable
Ultra Reflective Letter Label
For use in low temperature applications
Specifications
Autoclavable
None
Blank
Roll
Ink Jet
Non-sterile
4/Pk.
15 yd.
0.75 in.
1 in.
Rubber
Yellow
Matte
Single-side Printable
Ultra Reflective Letter Label
For use in low temperature applications
13.7 m x 19 mm
13.7 m
19 mm
2.54 cm
Product Suggestions
Product Suggestions