missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ Wrinkle Textured Latex Coated Cotton Gloves

Catalog No. 296367719
Change view
Click to view available options
Back Material:
Cotton
Size:
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Back Material Size
296367719 Cotton Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296367719 Supplier SHOWA™ Supplier No. L66NFW08

Gloves provide heavy duty protection with a natural latex coating for an enhanced grip in wet or dry conditions. The soft liner provides comfort and flexibility, while a reinforced safety cuff offers additional protection.

  • Heavy duty protection
  • Fully coated with natural latex
  • Texturized finish for enhanced grip
  • Cotton flannel liner increases comfort
  • Optional elasticated back for a secure fit
  • Reinforced safety cuff provides additional protection
  • In compliance 21 CFR 177.2600

Specifications

Back Material Cotton
Certifications/Compliance FDA 21 CFR 177.2600
Chemotherapeutic Rated No
Coated Area Palm, Fingers
Construction Seamless Knit
Cuff Material Reinforced Cotton
Coating Material Latex
Color Yellow
Exterior Finish Fully Textured
Hand Specific
Latex Free No
Material Cotton
Product Type Dip-coated Gloves
Size Small
For Use With (Application) Landscaping, Wood Cutting, Light Chemical Handling, and General Maintenance
Glove Cuff Color Natural
Glove Cuff Style Safety
Grip Design Rough
Liner Material Flannel
Resistant To Cuts, Chemicals, Abrasion, Cold
Series L66NFW
Show More Show Less