missing translation for 'onlineSavingsMsg'
Learn More

Sigma-Aldrich Wright Stain, Modified

Catalog No. 01288055
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01288055 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01288055 Supplier Sigma-Aldrich Supplier No. WS321L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For differential staining of the cellular elements of blood.

For dip, rack, and batch staining techniques.

  • Wright stain, 0.3%, buffered at pH 6.8 in methanol
  • Contains stabilizers and surfactant
  • Form: solution
  • Shelf life: expiry date on the label
  • IVD: for in vitro diagnostic use
  • Concentration: 0.3%
  • Techniques: microbe ID, suitable for staining
  • pH: 6.75 to 6.95 (25°C).
  • Applications: histology and hematology
  • Storage temperature: room temperature

Specifications

Quantity 1 L
Product Type Wright Stain