Learn More
Wright's Stain
CAS: 68988-92-1 | C36H27Br4N3O5S+2 | 933.304 g/mol
$119.86 - $337.74
Chemical Identifiers
| CAS | 68988-92-1 |
|---|---|
| Molecular Formula | C36H27Br4N3O5S+2 |
| Molecular Weight (g/mol) | 933.304 |
| MDL Number | MFCD00082143 |
| InChI Key | AXIKDPDWFVPGOD-UHFFFAOYSA-O |
| PubChem CID | 25113599 |
| IUPAC Name | [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;2-(2,4,5,7-tetrabromo-3,6-dihydroxyxanthen-10-ium-9-yl)benzoic acid |
| SMILES | CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.C1=CC=C(C(=C1)C2=C3C=C(C(=C(C3=[O+]C4=C(C(=C(C=C24)Br)O)Br)Br)O)Br)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1790314
|
Thermo Scientific Chemicals
A1790314 |
25 g |
Each for $119.86
|
|
|||||
|
AAA1790322
|
Thermo Scientific Chemicals
A1790322 |
100 g |
Each for $337.74
|
|
|||||
Description
Useful stain for blood and for bone marrow films
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 68988-92-1 | |
| 933.304 | |
| AXIKDPDWFVPGOD-UHFFFAOYSA-O | |
| [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;2-(2,4,5,7-tetrabromo-3,6-dihydroxyxanthen-10-ium-9-yl)benzoic acid |
| C36H27Br4N3O5S+2 | |
| MFCD00082143 | |
| 25113599 | |
| CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.C1=CC=C(C(=C1)C2=C3C=C(C(=C(C3=[O+]C4=C(C(=C(C=C24)Br)O)Br)Br)O)Br)C(=O)O |
Specifications
| 68988-92-1 | |
| MFCD00082143 | |
| CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.C1=CC=C(C(=C1)C2=C3C=C(C(=C(C3=[O+]C4=C(C(=C(C=C24)Br)O)Br)Br)O)Br)C(=O)O | |
| 933.304 | |
| 25 g |
| C36H27Br4N3O5S+2 | |
| AXIKDPDWFVPGOD-UHFFFAOYSA-O | |
| [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;2-(2,4,5,7-tetrabromo-3,6-dihydroxyxanthen-10-ium-9-yl)benzoic acid | |
| 25113599 | |
| Wright's Stain |
Safety and Handling
GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
EINECSNumber : 273-541-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only