missing translation for 'onlineSavingsMsg'
Learn More

Wright's Stain

Catalog No. AAA1790314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1790314 25 g
AAA1790322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1790314 Supplier Thermo Scientific Chemicals Supplier No. A1790314
Only null left

CAS: 68988-92-1 | C36H27Br4N3O5S+2 | 933.304 g/mol

Useful stain for blood and for bone marrow films

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68988-92-1
Molecular Formula C36H27Br4N3O5S+2
Molecular Weight (g/mol) 933.304
MDL Number MFCD00082143
InChI Key AXIKDPDWFVPGOD-UHFFFAOYSA-O
PubChem CID 25113599
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;2-(2,4,5,7-tetrabromo-3,6-dihydroxyxanthen-10-ium-9-yl)benzoic acid
SMILES CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.C1=CC=C(C(=C1)C2=C3C=C(C(=C(C3=[O+]C4=C(C(=C(C=C24)Br)O)Br)Br)O)Br)C(=O)O

Specifications

Quantity 25 g
Chemical Name or Material Wright's Stain
Hazard Category H302-H318
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
EINECSNumber 273-541-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only