missing translation for 'onlineSavingsMsg'
Learn More

Wright's Stain, Regular Formula, for Staining and General Differentiation of Blood Corpuscles, Ricca Chemical

Catalog No. 936016
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
936016 500 mL Amber Glass Bottle
936032 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 936016 Supplier Ricca Chemical Company Supplier No. 936016
Only null left

Staining and General Differentiation of Blood Corpuscles

Specifications

Chemical Name or Material Wright's Stain
Name Note Regular Formula
CAS 67-56-1, 68988-92-1, 7558-79-4, 7778-77-0, 7664-38-2, 7732-18-5
CAS Min % 97.0, 0.38, 0.08, 0.04, 0.0010
CAS Max % 100.0, 0.39, 0.09, 0.04, 0.0010
Grade Laboratory
Packaging Amber Glass Bottle
Color Dark Purple
pH 6.30 to 6.50
Quantity 500 mL
Physical Form Liquid
Show More Show Less