missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wright's Eosine Methylene Blue, For Microscopy, MilliporeSigma™
Specifications
Specifications
| Chemical Name or Material | Wright's Eosine Methylene Blue |
| Odor | Odorless |
| Packaging | Glass bottle |
| Color | Dark Green |
| Quantity | 25 g |
| Physical Form | Solid |