missing translation for 'onlineSavingsMsg'
Learn More

Sigma-Aldrich Wright-Giemsa Stain, Modified

Catalog No. 01288067
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01288067 1 L
01288049 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01288067 Supplier Sigma-Aldrich Supplier No. WG321L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

When blood films are stained using Wright-Giemsa stain, the white blood cell nucleus and cytoplasm will take on characteristic blue or pink coloration.

The combination of purified eosin and thiazine dyes in the product eliminates inconsistent staining and yields reproducible chromogenic responses.

  • Form: solution
  • Shelf Life: expiry date on the label
  • IVD: for in vitro diagnostic use
  • Concentration: 0.4% (w/v) in methanol (buffered at pH 6.8)
  • Applications: hematology and histology
  • Storage temperature: room temperature

Specifications

Quantity 1 L
Product Type Wright-Giemsa Stain