Learn More
GA International Wrap-Around Cryogenic Thermal-Transfer Labels 1.14"" x 0.59"" + 1.5"" (wrap) / 29mm x 15mm + 38.1mm (wrap) Rectangle White + Transparent From -196C to +100C (-321F to +212F)
Catalog No. 5000571541
Encompass
Self-laminating cryogenic labels that protect the printed information from alcohol, bleach, DMSO, and detergents to ensure the secure identification of tubes and cryo vials. Designed for long-term preservation under cryogenic laboratory conditions, including liquid nitrogen (-196C/-321F), -80C (-112F) freezers, and dry ice, and can withstand temperatures up to +100C (+212F). Labels can be written on with cryo markers, and printed with all major brands of thermal-transfer printers.
Catalog No. 5000571541 Supplier GA International Supplier No. HBTT346NPNOTC32WH
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Self-laminating cryogenic labels that protect the printed information from alcohol, bleach, DMSO, and detergents to ensure the secure identification of tubes and cryo vials. Designed for long-term preservation under cryogenic laboratory conditions, including liquid nitrogen (-196C/-321F), -80C (-112F) freezers, and dry ice, and can withstand temperatures up to +100C (+212F). Labels can be written on with cryo markers, and printed with all major brands of thermal-transfer printers.